missing translation for 'onlineSavingsMsg'
Learn More

TORC2 Antibody, Novus Biologicals™

Product Code. 18381973 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Conditionnement:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
18381973 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18381973 Fournisseur Novus Biologicals Code fournisseur NBP312449

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

TORC2 Polyclonal antibody specifically detects TORC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Spécification

Antigen TORC2
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2
Host Species Rabbit
Immunogen Phosphorylated synthetic peptide corresponding to unique amino acid sequence on TORC2 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 200186
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.