missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TORC1 Antibody (1B5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51631
This item is not returnable.
View return policy
Description
TORC1 Monoclonal specifically detects TORC1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| TORC1 | |
| Monoclonal | |
| Unconjugated | |
| CREB regulated transcription coactivator 1, FLJ14027WAMTP1, KIAA0616Transducer of regulated cAMP response element-binding protein 1, MECT1CREB-regulated transcription coactivator 1, mucoepidermoid carcinoma translocated 1, Mucoepidermoid carcinoma translocated protein 1, TORC1TORC-1, Transducer of CREB protein 1, transducer of regulated cAMP response element-binding protein (CREB) 1 | |
| Mouse | |
| 67 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 23373 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1B5 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| CRTC1 | |
| Purified recombinant fragment of human CRTC1 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction