missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR4A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA560865
This item is not returnable.
View return policy
Description
Immunogen sequence: SRLVLYPETS RKYRPRVEHR SRAQRCLLLL VAIVGFQVLN AIENLDDNAQ RYDLDGLEKA LQRAVFGQPA AVSRIVALMR D Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
TOR4A gene ontology annotations related to this gene include platelet degranulation.
Specifications
| TOR4A | |
| Polyclonal | |
| Unconjugated | |
| TOR4A | |
| C9H9orf167; C9orf167; RGD1308019; TOR4A; Torsin family 4 member A; torsin family 4, member A; torsin family protein C9orf167; torsin-4A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54863 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| Lot-Specific | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NXH8 | |
| TOR4A | |
| Recombinant protein corresponding to Human TOR4A | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction