missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOR3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119472
This item is not returnable.
View return policy
Description
TOR3A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The function remains unknown.
Specifications
| TOR3A | |
| Polyclonal | |
| Unconjugated | |
| TOR3A | |
| Adir; ADIR2; ATP-dependant interferon response protein 1; ATP-dependant interferon responsive; ATP-dependent interferon-responsive protein; si:ch211-198n5.8; Tor3a; Torsin family 3 member A; torsin family 3, member A; torsin-3A | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 30935, 64222 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9ER38, Q9H497 | |
| TOR3A | |
| Recombinant protein Torsin 3A. The antigen corresponds to amino acid range 125-374 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction