missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR1AIP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21459-1-AP-150UL
This item is not returnable.
View return policy
Description
Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).Specifications
| TOR1AIP1 | |
| Polyclonal | |
| Unconjugated | |
| TOR1AIP1 | |
| LAP1B, TOR1AIP1, torsin A interacting protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26092 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5JTV8 | |
| TOR1AIP1 | |
| TOR1AIP1 Fusion Protein Ag14823 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction