missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR/mTOR, p Ser2448 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 8 publications
Brand: Novus Biologicals NB600-607
This item is not returnable.
View return policy
Description
TOR/mTOR Polyclonal specifically detects TOR/mTOR in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TOR/mTOR (p Ser2448) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR | |
| Rabbit | |
| 288.89 kDa | |
| 0.1 mg | |
| Cancer, Macroautophagy, mTOR Pathway, Phospho Specific | |
| 2475 | |
| Human, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.1 mg/ml | |
| Western Blot 1:500-1:2000, ELISA 1:10000-1:100000, Immunohistochemistry 5.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, N/A | |
| P42345 | |
| MTOR | |
| TOR/TOR/mTOR [p Ser2448] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a phosphorylated synthetic peptide corresponding a c-terminal region near Serine 2448 of human TOR/mTOR. (Uniprot: P42345) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reactivity occurs with phosphorylated mTOR from human derived tissues and cells. Reactivity against mTOR from other species has not been determined, however, reactivity with mouse and rat is suggested based on protein sequence homologies. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction