missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOR/mTOR, p Ser2448 Antibody (1G6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26406-100ul
This item is not returnable.
View return policy
Description
TOR/mTOR Monoclonal antibody specifically detects TOR/mTOR in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| TOR/mTOR | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 1G6 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR | |
| A synthesized peptide derived from Human TOR/mTOR [p Ser2448] [UniProt P42345] | |
| 100 μL | |
| Autophagy, Cancer, Cell Cycle and Replication, DNA Repair, Hypoxia, Macroautophagy, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control | |
| 2475 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction