missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TopVision Agarose Tablets Promotion

Product Code. 15737546 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1000 Tablets (0.5 g each)
200 Tablets (0.5 g each)
Unit Size:
1000 tablets
200 tablets
This item is not returnable. View return policy

Product Code. 15737546

Brand: Thermo Scientific™ R2801

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality, multipurpose agarose tablets in blister packaging.

Description
TopVision Agarose Tablets, low EEO (LE), is multipurpose agarose in tablet format delivered in a convenient blister pack. The agarose tablets are made of standard melting point agarose that yields high resolution, sharp DNA bands with high clarity and low background. Its optimized gel strength enhances ease of gel processing and handling. Manufactured using an innovative Organic Solvent-Free manufacturing process that is greener and more environmentally friendly.
Features
• Convenient tablet format - no messy weighing required.
• Fast dissolving protocol.
•“Green”- manufactured with significantly reduced environmental impact, using an innovative organic solvent-free technology.
• Enhanced resolution and reduced background.
Applications
Ideal for routine DNA and RNA gel electrophoresis and blotting assays.
Environmentally responsible Green Solution
Unlike traditional agarose powder, which production relies heavily on the use of toxic organic solvents, TopVision Agarose Tablets are manufactured using an innovative process that completely eliminates dependency on organic solvents and significantly reduces environmental impact while creating a safer working environment.

Appearance:White to off-white powder
> Ash:≤ 0.5%
> CAS:9012-36-6
> EEO:≤ 0.13
> Gel Strength:≥ 1200 g/cm2 (1% Gel)
> Gelling Point:36°C ± 1.5°C (1.5% gel)
> Hazardous:No
> Impurities (DNAses, RNases and Proteases):None
> Melting Point:88°C ± 1.5°C (1.5% gel)
> Moisture:≤ 10%
> Shelf Life:24 Months
> Shipping Condition:Ambient
> Solubility:Clear colorless solution at 1g in 100 mL water
> Storage Condition:Ambient
> Sulfate:≤ 0.15%

TRUSTED_SUSTAINABILITY

Specifications

Product Type Agarose Tablets
For Use With (Application) For routine DNA and RNA gel electrophoresis and blotting assays
Quantity 200 Tablets (0.5 g each)
Storage Requirements Ambient
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.