missing translation for 'onlineSavingsMsg'
Learn More

Topotecan, MedChemExpress

Product Code. 16445848
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16445848

Brand: Medchem Express HY13768

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Topotecan (SKF 104864A; NSC 609669) is an orally active and potent Topoisomerase I inhibitor. Topotecan induces cell cycle arrest in G0/G1 and S phases and promotes apoptosis. Topotecan shows anticancer activity.

Topotecan (SKF 104864A

Specifications

Chemical Name or Material Topotecan
CAS 123948-87-8
Purity Grade Notes Research
Molecular Formula C23H23N3O5
Quantity 10 mg
Synonym SKF 104864A NSC 609669
SMILES O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=CC5=C(CN(C)C)C(O)=CC=C5N=C4C3=C2)=O
Molecular Weight (g/mol) 421.45
Formula Weight 421.45
Grade Research
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.