missing translation for 'onlineSavingsMsg'
Learn More

Topotecan Hydrochloride, Mikromol

Product Code. 30459370
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459370 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459370 Supplier Mikromol Supplier No. MM1330.000250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Topotecan Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Topotecan Hydrochloride
CAS 119413-54-6
Analyte or Component Names Topotecan Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C23 H23 N3 O5 . Cl H
Certifications/Compliance ISO 9001
Synonym (4S)-10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione Hydrochloride (1:1), 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-, monohydrochloride, (4S)- (9CI), 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-[(dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-, monohydrochloride, (S)-, (S)-10-[(Dimethylamino)methyl]-4-ethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[l,2-b]quinoline-3,14(4H,12H)-dione monohydrochloride, Hycamtin, NSC 609669, Nogitecan hydrochloride, SKF 104864A, SKFS 104864A, Topotecan chlorohydrate, Topotecan hydrochloride, Xinze, SKF-S-104864-A, SKF-S-104864, SKF-S-106567
InChI Formula InChI=1S/C23H23N3O5.ClH/c1-4-23(30)16-8-18-20-12(9-26(18)21(28)15(16)11-31-22(23)29)7-13-14(10-25(2)3)19(27)6-5-17(13)24-20;/h5-8,27,30H,4,9-11H2,1-3H3;1H/t23-;/m0./s1
Solution Type Neat
SMILES Cl.CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(CN(C)C)c(O)ccc5nc34)C2=O
Molecular Weight (g/mol) 457.91
Quantity 250 mg
Formula Weight 457.1404 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.