missing translation for 'onlineSavingsMsg'
Learn More

Topotecan-d6, TRC

Product Code. 30240530
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240530 1 mg 1mg
30240973 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240530 Supplier Toronto Research Chemicals Supplier No. TRCT5425021MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1044904-10-0
Molecular Formula C23 2H6 H17 N3 O5
Molecular Weight (g/mol) 427.48
Synonym 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 10-[[di(methyl-d3)amino]methyl]-4-ethyl-4,9-dihydroxy-, (4S)-, Topotecan-d6, Topotecan D6 (N,N-dimethyl D6)
SMILES [2H]C([2H])([2H])N(Cc1c(O)ccc2nc3C4=CC5=C(COC(=O)[C@]5(O)CC)C(=O)N4Cc3cc12)C([2H])([2H])[2H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C23H23N3O5/c1-4-23(30)16-8-18-20-12(9-26(18)21(28)15(16)11-31-22(23)29)7-13-14(10-25(2)3)19(27)6-5-17(13)24-20/h5-8,27,30H,4,9-11H2,1-3H3/t23-/m0/s1/i2D3,3D3
Formula Weight 427.2014
Chemical Name or Material Topotecan-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.