missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOPORS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35863-100ul
This item is not returnable.
View return policy
Description
TOPORS Polyclonal antibody specifically detects TOPORS in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TOPORS | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| EC 6.3.2.-, LUNtopoisomerase I binding, arginine/serine-rich, p53-binding protein 3, p53BP3, retinitis pigmentosa 31 (autosomal dominant), RP31, SUMO1-protein E3 ligase Topors, topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase, Topoisomerase I-binding arginine/serine-rich protein, Topoisomerase I-binding RING finger protein, TP53BPLE3 ubiquitin-protein ligase Topors, Tumor suppressor p53-binding protein 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of mouse TOPORS (NP_598858.2).,, Sequence:, PDSKCPICLDRFDNVSYLDRCLHKFCFRCVQEWSKNKAECPLCKQPFDSIFHSVRAEDDFKEYVLRPSYNGSFTNPEVRRFRYRTTMTRERSASLYSPSST | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10210 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction