missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOPORS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-56069-25ul
This item is not returnable.
View return policy
Description
TOPORS Polyclonal specifically detects TOPORS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| TOPORS | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL | |
| EC 6.3.2.-, LUNtopoisomerase I binding, arginine/serine-rich, p53-binding protein 3, p53BP3, retinitis pigmentosa 31 (autosomal dominant), RP31, SUMO1-protein E3 ligase Topors, topoisomerase I binding, arginine/serine-rich, E3 ubiquitin protein ligase, Topoisomerase I-binding arginine/serine-rich protein, Topoisomerase I-binding RING finger protein, TP53BPLE3 ubiquitin-protein ligase Topors, Tumor suppressor p53-binding protein 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| TOPORS | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GGATCQIQGVQTNDDLNNDSDDSSDNCVIVGFVKPLAERTPELVELSSDSEDLGSYEKMETVKTQEQEQSYSSGDSDVSRCSSPHSVLGKDEQINKGHCDSSTRIKSKKEEKRSTSLSSPRNL | |
| 25 μL | |
| Zinc Finger | |
| 10210 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction