missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Topo III beta 293T Cell Transient Overexpression Lysate
Shop All Bio Techne ProductsSpecifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Apoptosis, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors |
| Target | Topo III beta |
| Synonym | DNA topoisomerase 3-beta-1, DNA topoisomerase III beta-1, EC 5.99.1.2, FLJ39376, TOP3B1, topoisomerase (DNA) III beta, topoisomerase III beta |
| Gene Symbol | TOP3B |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?