missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOPAZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561516
This item is not returnable.
View return policy
Description
Immunogen sequence: RSASEELKLS CQRTIPMTGK RTWPYYSCAR ISAWCWKKAS LPESSYFLRG SQESCRQVDV PKHQTNQTHL TDSKLLLQSS LTETNTESSS KEKLDSNSNC LS Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 59%.
The testis and ovary specific PAZ domain containing 1 (TOPAZ1) protein is a highly conserved protein that is highly expressed in the gonads during germ cell meiosis, suggesting that it may play a role in germ cell development. It contains a PAZ domain and a zinc finger domain and is exclusively expressed in adult testis and fetal ovary.
Specifications
| TOPAZ1 | |
| Polyclonal | |
| Unconjugated | |
| TOPAZ1 | |
| C3orf77; Gm9524; Protein TOPAZ1; RGD1563398; testis and ovary specific PAZ domain containing 1; testis and ovary-specific PAZ domain gene 1; testis- and ovary-specific PAZ domain-containing protein 1; topaz 1; Topaz1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 375337 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N9V7 | |
| TOPAZ1 | |
| Recombinant protein corresponding to Human TOPAZ1. Recombinant protein control fragment (Product #RP-100229). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction