missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOP3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575799
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
DNA topoisomerases are nuclear enzymes that regulate the topological structure of DNA by transiently breaking and rejoining DNA strands. Although DNA topoisomerase I (Topo I) and Topo II relax both positive and negative supercoils, Topo III relaxes only negative supercoils. Topo IIIalpha exists as a long and a short isoform, which are produced by alternative splicing. Topo IIIalpha, which localizes to the nucleolus, is constitutively expressed and remains at high levels throughout the cell cycle in HL-60 cells. Topo IIIbeta exists as three isoforms,beta-1,beta -2 and beta-3, also produced by alternative splicing. Topo IIIbeta-1 is expressed in testis, heart, and skeletal muscle, whereas Topo IIIbeta-2 is expressed in thymus, kidney and pancreas.
Specifications
| TOP3A | |
| Polyclonal | |
| Unconjugated | |
| Top3a | |
| DNA topoisomerase 3-alpha; DNA topoisomerase III alpha; Top IIIa; TOP3; TOP3A; topo III-alpha; topoisomerase (DNA) 3 alpha; topoisomerase (DNA) III alpha; topoisomerase 3 alpha; ZGRF7; zinc finger, GRF-type containing 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7156 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.2 | |
| Q13472 | |
| Top3a | |
| Synthetic peptide corresponding to residues in Human TOP3A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction