missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554984
This item is not returnable.
View return policy
Description
Immunogen sequence: LWKEDLAAFV EELDKVESQE REDVLAGMSG KAIKGKVGKP KVKKLQLEET MPSPYGRRII PEITAMKADA SKKLLKKKKG DLDTAAVKVE FDEEFSGAPV EGAGEEALTP SVPINKGPKP KREKKEPGTR VRKTPTSSGK PSA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 40%.
TOP2B (DNA Topoisomerase II Beta) is a Protein Coding gene. This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This nuclear enzyme is involved in processes such as chromosome condensation, chromatid separation, and the relief of torsional stress that occurs during DNA transcription and replication. It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another, thus altering the topology of DNA. Two forms of this enzyme exist as likely products of a gene duplication event. The gene encoding this form, beta, is localized to chromosome 3 and the alpha form is localized to chromosome 17. The gene encoding this enzyme functions as the target for several anticancer agents and a variety of mutations in this gene have been associated with the development of drug resistance. Reduced activity of this enzyme may also play a role in ataxia-telangiectasia. Alternative splicing results in multiple transcript variants.
Specifications
| TOP2B | |
| Polyclonal | |
| Unconjugated | |
| TOP2B | |
| antigen MLAA-44; D230016L12Rik; DNA topoisomerase 2-beta; DNA topoisomerase II beta; DNA topoisomerase II, 180 kD; DNA topoisomerase II, beta isozyme; Top-2; Top2b; top2beta; TOPIIB; topo II beta; topoisomerase (DNA) II beta; topoisomerase (DNA) II beta 180kDa; topoisomerase II beta; topoisomerase IIb; U937 associated antigen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21974, 361100, 7155 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.40 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02880, Q64511 | |
| TOP2B | |
| Recombinant protein corresponding to Human TOP2B. Recombinant protein control fragment (Product #RP-92454). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction