missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2A Rabbit anti-Human, Clone: TOP2A/6570R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | TOP2A |
|---|---|
| Clone | TOP2A/6570R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
TOP2A Monoclonal specifically detects TOP2A in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| TOP2A | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 7153 | |
| IgG | |
| Protein A or G purified |
| TOP2A/6570R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A | |
| Recombinant human TOP2A fragment (aa1352-1493) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title