missing translation for 'onlineSavingsMsg'
Learn More

TOP2A Rabbit anti-Human, Clone: TOP2A/4397R, Novus Biologicals™

Product Code. 18362573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362573 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362573 Supplier Novus Biologicals Supplier No. NBP30773520UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TOP2A Monoclonal specifically detects TOP2A in Human samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP2A
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TOP2A/4397R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to TOP2A residues within aa1431-1531 of TOP2A
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7153
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.