missing translation for 'onlineSavingsMsg'
Learn More

TOP2A Antibody (TOP2A/1362), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18408013 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408013 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408013 Supplier Novus Biologicals Supplier No. NBP254545AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TOP2A Monoclonal antibody specifically detects TOP2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP2A
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone TOP2A/1362
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A
Host Species Mouse
Immunogen Recombinant fragment of human TOP2A (around aa1352-1493) (exact sequence is proprietary) (Uniprot: P11388)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7153
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.