missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TOP2A Monoclonal antibody specifically detects TOP2A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, CyTOF
Specifications
Specifications
| Antigen | TOP2A |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | TOP2A/1361 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | DNA topoisomerase (ATP-hydrolyzing), DNA topoisomerase 2-alpha, DNA topoisomerase II, 170 kD, DNA topoisomerase II, alpha isozyme, EC 5.99.1.3, TOP2DNA gyrase, topoisomerase (DNA) II alpha (170kD), topoisomerase (DNA) II alpha 170kDa, TP2A |
| Host Species | Mouse |
| Immunogen | Recombinant human TOP2A fragment (aa1352-1493) (Uniprot: P11388) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?