missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP1MT Antibody (rTOP1MT/488) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14279JF635
This item is not returnable.
View return policy
Description
TOP1MT Monoclonal antibody specifically detects TOP1MT in Human samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| TOP1MT | |
| Monoclonal | |
| Janelia Fluor 635 | |
| mitochondrial, mitochondrial DNA topoisomerase I, topoisomerase (DNA) I, mitochondrial | |
| Recombinant full-length human TOP1MT protein (Uniprot: Q969P6 ) | |
| 0.1 mL | |
| Cancer | |
| 116447 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| rTOP1MT/488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction