Learn More
Invitrogen™ TOP1 Recombinant Rabbit Monoclonal Antibody (SC69-03)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532228
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
Specifications
| TOP1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P11387, Q04750 | |
| Top1 | |
| Synthetic peptide within Human TOP1 aa 1-49. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ChIP Assay, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| SC69-03 | |
| Unconjugated | |
| Top1 | |
| Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; Topoisomerase 1; type I DNA topoisomerase | |
| Rabbit | |
| Protein A | |
| RUO | |
| 21969, 7150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.