missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOP1 Monoclonal Antibody (1291CT875.142.166)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537639

Product Code. 17137228

  • £455.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TOP1 Monoclonal Antibody for Western Blot, Flow

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TOP1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P11387
Top1
recombinant protein from the N-terminal region of human TOP1.
400 μL
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Western Blot
1291CT875.142.166
Unconjugated
Top1
Ab2-086; AI467334; D130064I21Rik; DNA topoisomerase 1; DNA topoisomerase I; RP3-511B24.2; TOP1; Top-1; TOPI; topoisomerase (DNA) I; Topoisomerase 1; type I DNA topoisomerase
Mouse
Protein G
RUO
7150
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.