Learn More
Stuart™ Top Plate Assembly
Stuart™ Top Plate Assembly for use with Stuart™ SD500 Digital Hotplate
Brand: Stuart™ 99953-11
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- Top Plate Assembly for use with Stuart™ Digital Hotplates
- Plate measures (300 x 500 mm)
Specifications
(300 x 500 mm) | |
Stuart™ Digital Hotplates (SD500) |