Learn More
Faster™ Top Box Kit for Decontamination of SafeFAST Elite Cabinets with 1.5 in. Camlock Outlet
Click to view available options
For Use With (Equipment):
SafeFAST Elite 109
SafeFAST Elite, Classic, Premium 218
SafeFAST Premium 209
SafeFAST Premium 212
Description
- Comes with a Camlock to attach fumigation machine
- Specific to SafeFAST Elite Cabinets
- Prevents fumigant escaping into the laboratory
Specifications
Specifications
| For Use With (Equipment) | SafeFAST Elite 109 |
| Width (Metric) | 200 mm |