Learn More
Faster™ Top Box Kit for Decontamination of SafeFAST Elite Cabinets with 1.5 in. Camlock Outlet
Product Code. 16218700
Click to view available options
For Use With (Equipment):
SafeFAST Elite 109
SafeFAST Elite, Classic, Premium 218
SafeFAST Premium 209
SafeFAST Premium 212
Product Code. 16218700
Brand: Faster™ FX0021547600
Top box attaches to the top of the Safety Cabinet to allow for fumigation and comes with a 1.5 in. Camlock.
  • Comes with a Camlock to attach fumigation machine
  • Specific to SafeFAST Elite Cabinets
  • Prevents fumigant escaping into the laboratory
Specifications
For Use With (Equipment) SafeFAST Elite 109
Width (Metric) 200 mm