Learn More
Faster™ Top Box Kit for Decontamination for SafeFAST Cabinets with 1 in. Camlock Outlet
Product Code. 16228700
Click to view available options
For Use With (Equipment):
SafeFAST Elite, Classic, Premium 209
SafeFAST Elite, Classic, Premium 212
SafeFAST Elite, Classic, Premium 215
SafeFAST Elite, Classic, Premium 218
Product Code. 16228700
Brand: Faster™ FX0021548300
Top box attaches to the top of the Safety Cabinet to allow for fumigation and comes with a 1 in. Camlock.
  • Comes with a Camlock to attach fumigation machine
  • Specific for SafeFAST Elite Cabinets
  • Prevents fumigant escaping into the laboratory
Specifications
For Use With (Equipment) SafeFAST Elite, Classic, Premium 215
Width (Metric) 200 mm