missing translation for 'onlineSavingsMsg'
Learn More

Toosendanin, TRC

Product Code. 30453176
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453176 10 mg 10mg
30449832 100 mg -
30449185 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453176 Supplier Toronto Research Chemicals Supplier No. TRCT54010010MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58812-37-6
Molecular Formula C30 H38 O11
Molecular Weight (g/mol) 574.62
Synonym [C(R),1α,3α,4β,5α,7α,12α,13α,14β,15β,17α]-3,12-Bis(acetyloxy)-14,15:21,23-diepoxy-1,7,19-trihydroxy-4,8-dimethyl-11-oxo-24-Norchola-20,22-diene-4-carboxaldehyde Cyclic 4,19-Hemiacetal, 14,15-Epoxy-4,10-(methanoxymethano)-1H-cyclopenta[a]phenanthrene, 24-Norchola-20,22-diene-4-carboxaldehyde Deriv., 28-Deacetylsendanin, Chuanliansu
SMILES CC(=O)O[C@@H]1C[C@H](O)[C@@]23CO[C@@H](O)[C@]1(C)[C@@H]2C[C@@H](O)[C@]4(C)[C@@H]3C(=O)[C@H](OC(=O)C)[C@@]5(C)[C@@H](C[C@H]6O[C@@]456)c7cocc7

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C30H38O11/c1-13(31)39-20-10-19(34)29-12-38-25(36)26(20,3)17(29)9-18(33)28(5)23(29)22(35)24(40-14(2)32)27(4)16(15-6-7-37-11-15)8-21-30(27,28)41-21/h6-7,11,16-21,23-25,33-34,36H,8-10,12H2,1-5H3/t16-,17-,18+,19-,20+,21+,23-,24-,25+,26+,27+,28+,29+,30+/m0/s1
Formula Weight 574.2414
Chemical Name or Material Toosendanin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.