missing translation for 'onlineSavingsMsg'
Learn More

TOOS, MedChemExpress

Product Code. 16471796
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471796 500 mg 500mg
16421589 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471796 Supplier Medchem Express Supplier No. HY15932
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TOOS, a Trinder's reagent, is a novel highly water-soluble aniline derivative; are widely used in diagnostic tests and biochemical tests.

TOOS, a Trinder's reagent, is a novel highly water-soluble aniline derivative

Specifications

Chemical Name or Material TOOS
CAS 82692-93-1
Color White
Purity Grade Notes Research
Molecular Formula C12H18NNaO4S
Quantity 500 mg
Synonym TOOS sodium salt
Solubility Information DMSO : ≥ 47 mg/mL (159.14 mM)
SMILES O=S(CC(O)CN(CC)C1=CC=CC(C)=C1)(O[Na])=O
Molecular Weight (g/mol) 295.33
Formula Weight 295.33
Percent Purity 99.76%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.