missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI M1, T11301, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Tool Steel, AISI M1, T11301, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Zirconium/Oxygen/Tantalum/Tin/Titanium/Aluminium/Arsenic/Niobium/Phosphorus/Nitrogen/Copper/Nickel/Cobalt/Manganese/Silicon/Carbon/Vanadium/Tungsten/Chromium/Molybdenum/Zinc/Boron/Sulfur/Lead/Antimony/Iron |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Zirconium 0.0013 %/Oxygen 0.0021 %/Tantalum 0.0024 %/Tin 0.0041 %/Titanium 0.0044 %/Aluminium 0.007 %/Arsenic 0.009 %/Niobium 0.013 %/Phosphorus 0.013 %/Nitrogen 0.037 %/Copper 0.091 %/Nickel 0.104 %/Cobalt 0.226 %/Manganese 0.318 %/Silicon 0.44 %/Carbon 0.85 %/Vanadium 1.19 %/Tungsten 1.98 %/Chromium 3.67 %/Molybdenum 8 %/Zinc (<0.003) %/Boron (0.0013) %/Sulfur (0.003) %/Lead (0.005) %/Antimony (0.006) %/Iron (83) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction