missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI D2, T30402, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Tool Steel, AISI D2, T30402, 100g
Specifications
Specifications
| Chemical Name or Material | Oxygen/Titanium/Niobium/Tin/Aluminium/Nitrogen/Cobalt/Phosphorus/Tungsten/Copper/Nickel/Manganese/Silicon/Molybdenum/Vanadium/Carbon/Chromium/Hydrogen/Magnesium/Bismuth/Selenium/Zinc/Antimony/Tantalum/Boron/Calcium/Lead/Zirconium/Arsenic |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Oxygen 0.003 %/Titanium 0.003 %/Niobium 0.004 %/Tin 0.005 %/Aluminium 0.014 %/Nitrogen 0.0152 %/Cobalt 0.02 %/Phosphorus 0.021 %/Tungsten 0.034 %/Copper 0.047 %/Nickel 0.114 %/Manganese 0.256 %/Silicon 0.256 %/Molybdenum 0.74 %/Vanadium 0.77 %/Carbon 1.519 %/Chromium 11.5 %/Hydrogen (<0.001) %/Magnesium (<0.001) %/Bismuth (<0.002) %/Selenium (<0.002) %/Zinc (<0.002) %/Antimony (<0.01) %/Tantalum (<0.01) %/Boron (0.0006) %/Calcium (0.0008) %/Lead (0.0008) %/Zirconium (0.002) %/Arsenic (0.01) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction