missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI A6, T30106, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Tool Steel, AISI A6, T30106, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Tin/Niobium/Nitrogen/Phosphorus/Tungsten/Vanadium/Cobalt/Copper/Nickel/Silicon/Carbon/Chromium/Molybdenum/Manganese/Boron/Oxygen/Aluminium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.003 %/Tin 0.004 %/Niobium 0.005 %/Nitrogen 0.0107 %/Phosphorus 0.012 %/Tungsten 0.013 %/Vanadium 0.014 %/Cobalt 0.015 %/Copper 0.05 %/Nickel 0.096 %/Silicon 0.39 %/Carbon 0.68 %/Chromium 1.04 %/Molybdenum 1.22 %/Manganese 1.98 %/Boron (0.001) %/Oxygen (0.0014) %/Aluminium (0.006) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction