missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI A6, T30106, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Tool Steel, AISI A6, T30106, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Niobium/Arsenic/Tin/Titanium/Nitrogen/Tungsten/Cobalt/Vanadium/Phosphorus/Aluminium/Copper/Nickel/Silicon/Carbon/Chromium/Molybdenum/Manganese/Calcium/Zirconium |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0009 %/Oxygen 0.0013 %/Niobium 0.003 %/Arsenic 0.008 %/Tin 0.008 %/Titanium 0.008 %/Nitrogen 0.0083 %/Tungsten 0.009 %/Cobalt 0.01 %/Vanadium 0.01 %/Phosphorus 0.014 %/Aluminium 0.019 %/Copper 0.142 %/Nickel 0.255 %/Silicon 0.32 %/Carbon 0.72 %/Chromium 0.99 %/Molybdenum 1.27 %/Manganese 1.91 %/Calcium (0.001) %/Zirconium (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction