missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI A2, T30102, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Tool Steel, AISI A2, T30102, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Titanium/Niobium/Tin/Nitrogen/Tungsten/Cobalt/Aluminium/Phosphorus/Copper/Nickel/Vanadium/Silicon/Manganese/Molybdenum/Carbon/Chromium/Lead/Calcium/Antimony/Zirconium/Arsenic |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.001 %/Oxygen 0.001 %/Titanium 0.0029 %/Niobium 0.004 %/Tin 0.005 %/Nitrogen 0.011 %/Tungsten 0.011 %/Cobalt 0.013 %/Aluminium 0.017 %/Phosphorus 0.019 %/Copper 0.077 %/Nickel 0.144 %/Vanadium 0.21 %/Silicon 0.28 %/Manganese 0.45 %/Molybdenum 0.97 %/Carbon 0.99 %/Chromium 4.99 %/Lead (0.0001) %/Calcium (0.001) %/Antimony (0.002) %/Zirconium (0.002) %/Arsenic (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction