missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI A2, T30102, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Tool Steel, AISI A2, T30102, 100g
Specifications
Specifications
| Chemical Name or Material | Titanium/Tin/Aluminium/Niobium/Nitrogen/Cobalt/Phosphorus/Copper/Nickel/Vanadium/Silicon/Manganese/Carbon/Molybdenum/Chromium/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Titanium 0.003 %/Tin 0.004 %/Aluminium 0.006 %/Niobium 0.006 %/Nitrogen 0.0096 %/Cobalt 0.014 %/Phosphorus 0.017 %/Copper 0.1 %/Nickel 0.14 %/Vanadium 0.22 %/Silicon 0.35 %/Manganese 0.54 %/Carbon 0.99 %/Molybdenum 1.01 %/Chromium 4.79 %/Tungsten (0.026) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction