Learn More
VICI Jour Chromatography™ ValvTool
Description
- For making connections in hard-to-reach areas
- The unique design with its slotted wrench allows tightening of nuts where a loop or a capillary might otherwise make it difficult.
- The ValvTool is ideal for Valco and Rheodyne™ valves and fittings, as well as most of the HPLC and PEEK fittings on the market with 1/4 in. and 5/16 in. heads
Specifications
Specifications
| Description | Ideal for nuts with 1/4 in. hex and 5/16 in. heads |