missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94377-0.02ml
This item is not returnable.
View return policy
Description
TOMM7 Polyclonal antibody specifically detects TOMM7 in Human samples. It is validated for Western Blot
Specifications
| TOMM7 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000 | |
| homolog of Tom7, mitochondrial import receptor subunit TOM7 homolog, TOM7, TOMM07,6.2 kd protein, Translocase of outer membrane 7 kDa subunit homolog, translocase of outer mitochondrial membrane 7 homolog (yeast) | |
| A synthetic peptide corresponding to a sequence within amino acids 1-55 of human TOMM7 (NP_061932.1). KRGADPGMPEPTVLSLLWG | |
| 0.02 mL | |
| Endocrinology | |
| 54543 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction