missing translation for 'onlineSavingsMsg'
Learn More

TOMM6 Antibody, Novus Biologicals™

Product Code. 18319831 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319831 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319831 Supplier Novus Biologicals Supplier No. NBP312284

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TOMM6 Polyclonal antibody specifically detects TOMM6 in Human, Mouse, Rat, Bovine, Primate samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM6
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000, Immunoprecipitation
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias FLJ32622, mitochondrial import receptor subunit TOM6 homolog, over-expressed breast tumor protein, TOM6, translocase of outer membrane 6 kDa subunit homolog, translocase of outer mitochondrial membrane 6 homolog (yeast)
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 24-74 on human Mitochondrial import receptor subunit TOM6 homolog.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 100188893
Target Species Human, Mouse, Rat, Bovine, Primate
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.