missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA561807
This item is not returnable.
View return policy
Description
Immunogen sequence: RIEGLAPKLD PEEMKRKMRE DVISSIRNFL IYVALLRVTP FILKKLD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Specifications
| TOMM5 | |
| Polyclonal | |
| Unconjugated | |
| TOMM5 | |
| TOMM5 | |
| Recombit protein corresponding to Human TOMM5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N4H5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 401505 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction