missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM40 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18409-1-AP-20UL
This item is not returnable.
View return policy
Description
TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.Specifications
| TOMM40 | |
| Polyclonal | |
| Unconjugated | |
| TOMM40 | |
| C19orf1, D19S1177E, p38.5, PER EC1, PEREC1, Protein Haymaker, TOM40, TOMM40 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10452, 308416, 53333 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O96008, Q75Q40, Q9QYA2 | |
| Tomm40 | |
| TOMM40 Fusion Protein Ag13065 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction