missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOMM40 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578325
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Pig (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
Specifications
| TOMM40 | |
| Polyclonal | |
| Unconjugated | |
| TOMM40 | |
| AW539759; C19orf1; D19S1177E; haymaker protein; mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; mitochondrial outer membrane protein 35; mitochondrial outer membrane protein of 35 kDa; Mitochondrial outer membrane protein of 38 kDa; mitochondrial translocase of outer membrane protein; Mom35; OM38; outer membrane protein 38 kDa; p38.5; PER EC1; PEREC1; PER-EC1; Protein Haymaker; TOM40; Tom40a; Tomm40; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40; translocase of outer mitochondrial membrane 40 homolog; translocase of outer mitochondrial membrane 40 homolog (yeast) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10452 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.94 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O96008 | |
| TOMM40 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Human TOMM40. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction