missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOMM40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100084
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TOM40.
TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
Specifications
| TOMM40 | |
| Polyclonal | |
| Unconjugated | |
| TOMM40 | |
| AW539759; C19orf1; D19S1177E; haymaker protein; mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; mitochondrial outer membrane protein 35; mitochondrial outer membrane protein of 35 kDa; Mitochondrial outer membrane protein of 38 kDa; mitochondrial translocase of outer membrane protein; Mom35; OM38; outer membrane protein 38 kDa; p38.5; PER EC1; PEREC1; PER-EC1; Protein Haymaker; TOM40; Tom40a; Tomm40; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40; translocase of outer mitochondrial membrane 40 homolog; translocase of outer mitochondrial membrane 40 homolog (yeast) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10452, 308416, 53333 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O96008, Q75Q40, Q9QYA2 | |
| TOMM40 | |
| A synthesized peptide derived from human TOMM40(Accession O96008), corresponding to amino acid residues R173-Q223. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction