missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM34 Antibody (OTI2A9), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74550PECY7
This item is not returnable.
View return policy
Description
TOMM34 Monoclonal specifically detects TOMM34 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| TOMM34 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| TOMM34 | |
| Full length recombinant protein of human TOMM34 (NP_006800) produced in HEK293 cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry | |
| OTI2A9 | |
| Flow Cytometry | |
| hTom34, HTOM34P, mitochondrial import receptor subunit TOM34, outer mitochondrial membrane translocase (34kD), TOM34, Translocase of outer membrane 34 kDa subunit, translocase of outer mitochondrial membrane 34, URCC3 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10953.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction