missing translation for 'onlineSavingsMsg'
Learn More

TOMM22 Antibody (7C11), Novus Biologicals™

Product Code. 18375053
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375053 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375053 Supplier Bio-Techne Supplier No. NBP315219

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TOMM22 Monoclonal antibody specifically detects TOMM22 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOMM22
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 7C11
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias 1C9-2, hTom22, mitochondrial import receptor subunit TOM22 homolog, mitochondrial import receptor Tom22, MST065, TOM22MSTP065, Translocase of outer membrane 22 kDa subunit homolog, translocase of outer mitochondrial membrane 22 homolog (yeast)
Host Species Rabbit
Immunogen A synthesized peptide derived from human TOMM22. UniProt ID (Q9NS69).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 56993
Target Species Human
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.