missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOMM20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578300
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The mitochondrial preprotein translocases of the outer membrane (Tom) is a multisubunit protein complex that facilitates the import of nucleus-encoded precursor proteins across the mitochondrial outer membrane. The Tom machinery consists of import receptors for the initial binding of cytosolically synthesized preproteins and a general import pore (GIP) for the membrane translocation of various preproteins into the mitochondria. The import receptors include Tom20 and Tom22, which form a heteromeric receptor complex that initiates the insertion of newly synthesized proteins into the outer membrane and then directs the precursor protein into the GIP. In yeast, Tom22 is the essential component of the import receptor complex as it functions as both a receptor for the preproteins and serves as a docking point for both Tom20 and the GIP.
Specifications
| TOMM20 | |
| Polyclonal | |
| Unconjugated | |
| TOMM20 | |
| 1810060K07Rik; BB284719; Gm19268; KIAA0016; MAS20; mitochondrial 20 kDa outer membrane protein; Mitochondrial import receptor subunit TOM20 homolog; mKIAA0016; MOM19; outer mitochondrial membrane receptor rTOM20; outer mitochondrial membrane receptor Tom20; TOM20; TOMM20; translocase of outer mitochondrial membrane 20; translocase of outer mitochondrial membrane 20 homolog (yeast); translocase of outer mitochondrial membrane 20 homolog type II | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 266601, 67952, 9804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.22 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q15388, Q62760, Q9DCC8 | |
| TOMM20 | |
| Full length Human TOMM20 recombinant protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction