missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOMM20 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94821-0.02ml
This item is not returnable.
View return policy
Description
TOMM20 Polyclonal antibody specifically detects TOMM20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Gene Knock-Out
Specifications
| TOMM20 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin, Knockout Validated | |
| KIAA0016Outer mitochondrial membrane receptor Tom20, MAS20, MGC117367, Mitochondrial 20 kDa outer membrane protein, mitochondrial import receptor subunit TOM20 homolog, MOM19, TOM20, translocase of outer mitochondrial membrane 20 homolog (yeast), translocase of outer mitochondrial membrane 20 homolog type II | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-145 of human TOMM20 (NP_055580.1). YCIYFDRKRRSDPNFKNRLRERRKKQKLAKERAGLSKLPDLKDAEAVQKFFLEEIQLGEELLAQGEYEKGVDHLTNAIAVCGQPQQLLQVLQQTLPPPVFQMLLTKLPTISQRIVSAQSLAEDDVE | |
| 0.02 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 9804 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Gene Knock-Out | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction