missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOM70 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12828812
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828812 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828812 Supplier Invitrogen™ Supplier No. PA520664

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-20664 can be used with blocking peptide PEP-0784.

Functional mitochondria require up to 1000 proteins to function properly, with 99% synthesized as precursors in the cytoplasm and transported into the mitochondria with the aid of cytosolic chaperones and mitochondrial translocators (import components). Proteins to be imported are chaperoned to the mitochondria by the cytosolic heat shock protein (cHSP70) and are immediately pursued by Translocators of the Outer Membrane (TOMs), followed by transient interactions of the unfolded proteins with Translocators of the Inner Membrane (TIMs). TOMM70A is ubiquitously expressed in human tissues and localizes in the mitochondria. TOMM70A could play a significant role in the import of nuclear-encoded mitochondrial proteins with internal targeting sites such as ADP/ATP carriers and the uncoupling proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOM70
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TOMM70
Gene Accession No. O94826, Q75Q39, Q9CZW5
Gene Alias 2610044B22Rik; D16Ium22; D16Ium22e; D16Wsu109e; FLJ90470; KIAA0719; Mitochondrial import receptor subunit TOM70; mitochondrial precursor proteins import receptor; mKIAA0719; Tom70; Tomm70; Tomm70a; Translocase of outer membrane 70 kDa subunit; translocase of outer mitochondrial membrane 70; translocase of outer mitochondrial membrane 70 homolog A; translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae); translocase of outer mitochondrial membrane 70 homolog A (yeast); translocase of outer mitochondrial membrane protein 70
Gene Symbols TOMM70, Tomm70a
Host Species Rabbit
Immunogen A 17 amino acid peptide near the amino terminus of human TOM70.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28185, 304017, 9868
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.