missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOM70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA146188
This item is not returnable.
View return policy
Description
Suggested positive control: purified mitochondrial extracts and HeLa whole cell extract.
Functional mitochondria require up to 1000 proteins to function properly, with 99% synthesized as precursors in the cytoplasm and transported into the mitochondria with the aid of cytosolic chaperones and mitochondrial translocators (import components). Proteins to be imported are chaperoned to the mitochondria by the cytosolic heat shock protein (cHSP70) and are immediately pursued by Translocators of the Outer Membrane (TOMs), followed by transient interactions of the unfolded proteins with Translocators of the Inner Membrane (TIMs). TOMM70A is ubiquitously expressed in human tissues and localizes in the mitochondria. TOMM70A could play a significant role in the import of nuclear-encoded mitochondrial proteins with internal targeting sites such as ADP/ATP carriers and the uncoupling proteins.
Specifications
| TOM70 | |
| Polyclonal | |
| Unconjugated | |
| TOMM70 | |
| 2610044B22Rik; D16Ium22; D16Ium22e; D16Wsu109e; FLJ90470; KIAA0719; Mitochondrial import receptor subunit TOM70; mitochondrial precursor proteins import receptor; mKIAA0719; Tom70; Tomm70; Tomm70a; Translocase of outer membrane 70 kDa subunit; translocase of outer mitochondrial membrane 70; translocase of outer mitochondrial membrane 70 homolog A; translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae); translocase of outer mitochondrial membrane 70 homolog A (yeast); translocase of outer mitochondrial membrane protein 70 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100352274, 28185, 304017, 9868 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| O94826, Q75Q39, Q9CZW5 | |
| TOMM70, Tomm70a | |
| A synthetic peptide made to a region within residues 1-100 of the human TOM70 protein. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rabbit, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction