missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOM1L1 Mouse anti-Human, Clone: TOM1L1/4690, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07136-100ug
This item is not returnable.
View return policy
Description
TOM1L1 Monoclonal specifically detects TOM1L1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| TOM1L1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| OK/KNS-CL.3, Src-activating and signaling molecule protein, SRCASMSrc activating and signaling molecule, target of myb 1-like 1, target of myb1 (chicken) homolog-like 1, target of myb1 (chicken)-like 1, target of myb1-like 1, Target of Myb-like protein 1, TOM1-like protein 1 | |
| Recombinant full-length human TOM1L1 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| TOM1L1/4690 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10040 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction